Learn More
BIOHIPPO Human MYCBP Microsample
SDP
Catalog No. 5000314282
Encompass
Human MYCBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYCBP in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYCBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYCBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYCBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14282 Supplier BIOHIPPO Supplier No. BHE15211752
Human MYCBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYCBP in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYCBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYCBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYCBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color