Learn More
BIOHIPPO Human MyD88 Microsample
SDP
Catalog No. 5000314222
Encompass
Human MyD88 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MyD88 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MyD88 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MyD88 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MyD88 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14222 Supplier BIOHIPPO Supplier No. BHE15212101
Human MyD88 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MyD88 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MyD88 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MyD88 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MyD88 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color