Learn More
Biomatik Corporation Human Myelin Basic Protein (MBP) ELISA Kit
Supplier:  Biomatik Corporation EKU11301
Encompass_Preferred
Human Myelin Basic Protein (MBP) ELISA Kit for detecting MBP in serum, plasma, tissue homogenates, cell lysates, cerebrospinal fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 246.9-20000pg/ml. Sensitivity: 85.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02686. Gene ID: 4155. Target Synonyms: Myelin A1 protein; Myelin membrane encephalitogenic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8172