Learn More
Biomatik Corporation Human Myeloid Progenitor Inhibitory Factor 1 (MPIF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09674
Encompass_Preferred
Human Myeloid Progenitor Inhibitory Factor 1 (MPIF1) CLIA Kit for detecting MPIF1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.11pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P55773. Gene ID: 6368. Target Synonyms: CCL23; SCYA23; Ckb-8; MIP-3; CKb8; Chemokine (C-C-Motif) ligand 23; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 23; Macrophage inflammatory protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8469