Learn More
Biomatik Corporation Human Myeloperoxidase (MPO) CLIA Kit
Supplier:  Biomatik Corporation EKU08902
Encompass_Preferred
Human Myeloperoxidase (MPO) CLIA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05164. Gene ID: 4353. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8598