Learn More
BIOHIPPO Human MYH7B Microsample
SDP
Catalog No. 5000315060
Encompass
Human MYH7B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH7B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 074 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYH7B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYH7B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYH7B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15060 Supplier BIOHIPPO Supplier No. BHE15210962
Add to Cart
Add to Cart
Human MYH7B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH7B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 074 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYH7B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYH7B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYH7B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color