Learn More
BIOHIPPO Human MYL9 Microsample
SDP
Catalog No. 5000313881
Encompass
Human MYL9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYL9 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYL9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYL9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYL9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13881 Supplier BIOHIPPO Supplier No. BHE15211603
Human MYL9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYL9 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MYL9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MYL9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MYL9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color