Learn More
FUJIFILM Biosciences Human Myostatin Propeptide, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Catalog No. 50990676
Encompass_Preferred
Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and negative regulator of skeletal muscle mass. The recombinant myostatin propeptide binds and inhibits myostatin to enhance the repair and regeneration of injured muscle and bone.
Catalog No. 50-990-676 Supplier FUJIFILM Biosciences Supplier No. 1001421MG
Add to Cart
Add to Cart
Request a Quote
Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and negative regulator of skeletal muscle mass. The recombinant myostatin propeptide binds and inhibits myostatin to enhance the repair and regeneration of injured muscle and bone.