Learn More
FUJIFILM BIOSCIENCES INC Human Myostatin Propeptide, Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001425UG
Encompass_Preferred
Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and negative regulator of skeletal muscle mass. The recombinant myostatin propeptide binds and inhibits myostatin to enhance the repair and regeneration of injured muscle and bone.
Catalog No. 50-990-673