Learn More
Biomatik Corporation Human Myoglobin (MYO) CLIA Kit
Supplier:  Biomatik Corporation EKU09461
Encompass_Preferred
Human Myoglobin (MYO) CLIA Kit for detecting MYO in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 27.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02144. Gene ID: 4151. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8515