Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Myoglobin Recombinant Protein

Catalog No. PIRP75665
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75665 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75665 Supplier Invitrogen™ Supplier No. RP75665
Only null left
Add to Cart
Add to Cart

Recombinant Protein

RP-75665 contains purified recombinant human Myoglobin protein with an N-terminal hexahistidine tag. RP-75665 may be used as a positive control in many immunological assays.

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02144
Concentration 0.16 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 250 mM NaCl, 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 4151
Name Human Myoglobin
pH Range 8
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB
Common Name Myoglobin
Gene Symbol MB
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1- Gly154 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.