Learn More
Bioworld HUMAN MYOGLOBIN (RECOMBINANT)
Supplier:  Bioworld 220604241
Encompass_Preferred
Recombinant Human Myoglobin Source: E.Coli Background: Myoglobin is a cytoplasmic hemoprotein, expressed solely in cardiac myocytes and oxidative skeletal muscle fibers, that reversibly binds O(2) by its heme residue, a porphyrin rin...
Catalog No. 50-253-521