Learn More
Biomatik Corporation Human Myopalladin (MYPN) ELISA Kit
Supplier:  Biomatik Corporation EKU06084
Encompass_Preferred
Human Myopalladin (MYPN) ELISA Kit for detecting MYPN in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86TC9. Gene ID: 84665. Target Synonyms: MYOP; Sarcomeric Protein Myopalladin,145 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6282