Learn More
Biomatik Corporation Human Myosin Heavy Chain 11, Smooth Muscle (MYH11) ELISA Kit
Supplier:  Biomatik Corporation EKU06090
Encompass_Preferred
Human Myosin Heavy Chain 11, Smooth Muscle (MYH11) ELISA Kit for detecting MYH11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35749. Gene ID: 4629. Target Synonyms: AAT4; FAA4; SMHC; SMMHC; Myosin heavy chain, smooth muscle isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6357