Learn More
Biomatik Corporation Human Myosin Heavy Chain 4, Skeletal Muscle (MYH4) ELISA Kit
Supplier:  Biomatik Corporation EKU06094
Encompass_Preferred
Human Myosin Heavy Chain 4, Skeletal Muscle (MYH4) ELISA Kit for detecting MYH4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y623. Gene ID: 4622. Target Synonyms: MYH2B; Myosin heavy chain 2b; Myosin heavy chain, skeletal muscle, fetal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6624