Learn More
Biomatik Corporation Human Myosin Heavy Chain 8, Skeletal Muscle, Perinatal (MYH8) ELISA Kit
Supplier:  Biomatik Corporation EKU06102
Encompass_Preferred
Human Myosin Heavy Chain 8, Skeletal Muscle, Perinatal (MYH8) ELISA Kit for detecting MYH8 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13535. Gene ID: 4626. Target Synonyms: MyHC-peri; MyHC-pn; MyHC-perinatal; Myosin heavy chain, skeletal muscle, perinatal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8855