Learn More
Biomatik Corporation Human Myosin Light Chain Kinase 2 (MYLK2) ELISA Kit
Supplier:  Biomatik Corporation EKU12433
Encompass_Preferred
Human Myosin Light Chain Kinase 2 (MYLK2) ELISA Kit for detecting MYLK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H1R3. Gene ID: 85366. Target Synonyms: KMLC; skMLCK; MLCK2; Skeletal Muscle Myosin Light Chain Kinase; Myosin light chain kinase 2, skeletal/cardiac muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7167