Learn More
Sino Biological Anti-MZB1/PERP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15682RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PERP1 / MZB1 (rh MZB1; Catalog#15682-H08H; Q8WU39-1; Met 1- Thr 185). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4091