Learn More
Abcam Human N-368 Tau ELISA Kit- Intracellular, 1 x 96 Tests
Catalog No. 502668951
Encompass_Preferred
Human N-368 Tau ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human N-368 Tau with a sensitivity of 33.085 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-8951 Supplier Abcam Supplier No. AB3150591X96TESTS
Human N-368 Tau ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human N-368 Tau with a sensitivity of 33.085 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement