Learn More
Sino Biological Human NAA10 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14293MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ARD1 (rh ARD1; Catalog#14293-H20B; P41227; Met1-Ser235). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1599