Learn More
Biomatik Corporation Human N-Acetyltransferase 1 (NAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU06135
Encompass_Preferred
Human N-Acetyltransferase 1 (NAT1) ELISA Kit for detecting NAT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18440. Gene ID: 9. Target Synonyms: AAC1; Arylamine N-Acetyltransferase; Arylamide acetylase 1; Monomorphic arylamine N-acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4344