Learn More
Biomatik Corporation Human Na-Cl Cotransporter (NCCT) ELISA Kit
Supplier:  Biomatik Corporation EKU06139
Encompass_Preferred
Human Na-Cl Cotransporter (NCCT) ELISA Kit for detecting NCCT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55017. Gene ID: 6559. Target Synonyms: SLC12A3; TSC; NCC; Solute Carrier Family 12 Member 3,Sodium/Chloride Transporters; Thiazide-sensitive sodium-chloride cotransporter; Na-Cl symporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4404