Learn More
Biomatik Corporation Human N-Acylethanolamine Acid Amidase (NAAA) ELISA Kit
Supplier:  Biomatik Corporation EKU06140
Encompass_Preferred
Human N-Acylethanolamine Acid Amidase (NAAA) ELISA Kit for detecting NAAA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02083. Gene ID: 27163. Target Synonyms: ASAHL; PLT; N-Acylethanolamine-Hydrolyzing Acid Amidase; N-Acylsphingosine Amidohydrolase Like; Acid ceramidase-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6138