Learn More
Biomatik Corporation Human N-Acylsphingosine Amidohydrolase 2 (ASAH2) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12232
Encompass_Preferred
Human N-Acylsphingosine Amidohydrolase 2 (ASAH2) High-sensitive ELISA Kit for detecting ASAH2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NR71. Gene ID: 56624. Target Synonyms: HNAC1; non-lysosomal ceramidase; Neutral ceramidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5267