Learn More
Biomatik Corporation Human NAD Kinase (NADK) ELISA Kit
Supplier:  Biomatik Corporation EKU06142
Encompass_Preferred
Human NAD Kinase (NADK) ELISA Kit for detecting NADK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95544. Gene ID: 65220. Target Synonyms: Poly (P) /ATP NAD kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6215