Learn More
Biomatik Corporation Human NADH Dehydrogenase 5 (ND5) ELISA Kit
Supplier:  Biomatik Corporation EKU06145
Encompass_Preferred
Human NADH Dehydrogenase 5 (ND5) ELISA Kit for detecting ND5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.50-4000pg/ml. Sensitivity: 27.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P03915. Gene ID: 4540. Target Synonyms: MT-ND5; MTND5; NAD5; NADH-Ubiquinone Oxidoreductase Chain 5; Mitochondrially Encoded NADH Dehydrogenase 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5353