Learn More
Bioworld HUMAN NAFARELIN (RECOMBINANT)
Supplier:  Bioworld 220604271
Encompass_Preferred
Nafarelin acetate Source:E.Coli Background: Nafarelin acetate is a synthetic analog of the naturally occurring gonadotropin-releasing hormone (GnRH). Nafarelin stimulates the release of the pituitary gonadotropins, LH and FSH, result...
Catalog No. 50-253-524