Learn More
STEMCELL Technologies EasySep™ Human Naïve B Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1465970
Encompass
The EasySep™ Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep™ Human Naïve B Cell Enrichment Kit (Catalog #19254) for even faster cell isolations.
Catalog No. NC1465970 Supplier STEMCELL Technologies Supplier No. 17254
The EasySep™ Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep™ Human Naïve B Cell Enrichment Kit (Catalog #19254) for even faster cell isolations.