Learn More
Biomatik Corporation Human Na-K-Cl Cotransporter 2 (NKCC2) ELISA Kit
Supplier:  Biomatik Corporation EKU06150
Encompass_Preferred
Human Na-K-Cl Cotransporter 2 (NKCC2) ELISA Kit for detecting NKCC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13621. Gene ID: 6557. Target Synonyms: SLC12A1; NKCC2; Solute Carrier Family 12 Member 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; Kidney-specific Na-K-Cl symporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5296