Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NALP1 Synthetic Peptide

Catalog No. PEP0308 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0308 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0308 Supplier Invitrogen™ Supplier No. PEP0308
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 13 amino acids near the carboxy terminus of human NALP1. PEP-0308 can be used as a blocking peptide with polyclonal antibody PA5-20005.

NALP1 is a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 22861
Species Human
Purification Method purified
Accession Number Q9C000
Common Name NALP1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias CARD7; caspase recruitment domain protein; caspase recruitment domain protein 7; caspase recruitment domain-containing protein 7; CIDED; CLR17.1; death effector filament-forming Ced-4-like apoptosis protein; DEFCAP; DEFCAP-L/S; DKFZp586O1822; Gm14; Gm15; KIAA0926; NAC; nacht; NACHT, leucine rich repeat and PYD (pyrin domain) containing 1; NACHT, leucine rich repeat and PYD containing 1; NACHT, LRR and PYD containing protein 1; NACHT, LRR and PYD domains-containing protein 1; NACHT, LRR and PYD domains-containing protein 1a; NACHT/LRR/pyrin domain-containing protein 1; Nalp1; Nalp1a; NLR family protein 1; NLR family pyrin domain containing 1; NLR family, pyrin domain containing 1; NLR family, pyrin domain containing 1A; Nlrp1; Nlrp1a; Nucleotide-binding domain and caspase recruitment domain; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 1; PP1044; resistant anthrax lethal toxin; SLEV1; VAMAS1
Gene Symbol NLRP1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.