Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NALP2 Synthetic Peptide

Catalog No. PEP0348 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0348 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0348 Supplier Invitrogen™ Supplier No. PEP0348
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 12 amino acids near the carboxy terminus of human NALP2. PEP-0348 can be used as a blocking peptide with polyclonal antibody PA5-20213.

NALP2 belongs to a family of cytoplasmic proteins that have been implicated in cell responses to apoptotic and inflammatory stimuli. Unlike the prototypical NALP protein NALP1, NALP2 only contains a NACHT domain, leucine rich repeat (LRR), and pyrin-containing domain (PYD). This protein interacts with the adapter protein ASC in addition to CARD8 and caspase-1 to form an inflammasome with high proIL-1-b-processing activity and is thought to function as a modulator of NF-kappa-B and procaspase-1 activation in macrophages. It has also been suggested that NALP2, in addition to other NALP family members, can act as innate sensors of pathogens in a manner similar to the toll-like receptors (TLRs). At least two alternatively spliced transcript variants encoding distinct isoforms have been found for NALP2.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55655
Species Human
Purification Method purified
Accession Number Q9NX02
Common Name NALP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias CLR19.9; E330007A02Rik; FLJ20510; LOC684545; NACHT, leucine rich repeat and PYD containing 2; NACHT, LRR and PYD containing protein 2; NACHT, LRR and PYD domains-containing protein 2; NALP2; NBS1; NLR family pyrin domain containing 2; NLR family, pyrin domain containing 2; NLRP2; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 2; nucleotide-binding site protein 1; PAN1; PYPAF2; PYRIN domain and NACHT domain-containing protein 1; PYRIN-containing APAF1-like protein 2; similar to NACHT, leucine rich repeat and PYD containing 2
Gene Symbol NLRP2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.