Learn More
Sino Biological Anti-Alpha SNAP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14800RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SNAP-alpha (rh SNAP-alpha; Catalog#14800-H07E; NP_003818.2; Met1-Arg295). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1046