Learn More
Sino Biological Human NAPA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40010MM0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SNAP-alpha (rh SNAP-alpha; Catalog#14800-H07E; NP_003818.2; Met1-Arg295). SNAP-alpha specific IgG was purified by Human SNAP-alpha affinity chromatography.
Catalog No. 50-237-4076