Learn More
Biomatik Corporation Human Nardilysin (NRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU12057
Encompass_Preferred
Human Nardilysin (NRD1) ELISA Kit for detecting NRD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43847. Gene ID: 4898. Target Synonyms: NRD-C; N-Arginine Dibasic Convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8044