Learn More
Biomatik Corporation Human Natriuretic Peptide Receptor 1 (NPR1) ELISA Kit
Supplier:  Biomatik Corporation EKU06152
Encompass_Preferred
Human Natriuretic Peptide Receptor 1 (NPR1) ELISA Kit for detecting NPR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16066. Gene ID: 4881. Target Synonyms: ANPRA; ANPa; GUC2A; GUCY2A; NPR-A; Guanylate Cyclase A; Atrionatriuretic Peptide Receptor A; Atrial natriuretic peptide receptor type A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7135