Learn More
Biomatik Corporation Human Natural Cytotoxicity Triggering Receptor 3 (NCR3) ELISA Kit
Supplier:  Biomatik Corporation EKU11306
Encompass_Preferred
Human Natural Cytotoxicity Triggering Receptor 3 (NCR3) ELISA Kit for detecting NCR3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14931. Gene ID: 259197. Target Synonyms: CD337; 1C7; LY117; NKp30; Lymphocyte Antigen 117; Activating natural killer receptor p30; Natural killer cell p30-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6127