Learn More
iQ Biosciences Human Natural T Regulatory Cells, 1M cells/vial
Supplier:  iQ Biosciences IQBHU1NTR1
Encompass
High-quality, high-viability (typically > 80%) natural human T regulatory cells isolated from PBMCs with confirmed suppressive capacity. Used for a wide variety of in-vitro applications, including co-culture suppression and cytotoxicity assays. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1923856