Learn More
Sino Biological Human NBAS PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11707RP0120
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human NBAS fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4037