Learn More
Sino Biological Anti-NBL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10169MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NBL1 / DAND1 (rh NBL1 / DAND1; Catalog#10169-H08H; NP_005371.1; Met1-Asp180). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9578