Learn More
Sino Biological Human CD8B / P37 / LEU2 Protein 1mg
Supplier:  Sino Biological 11031HCCH1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD8B (P10966-1) (Met1-Pro170) was expressed with six amino acids (LEVLFQ) at the C- terminus.
Catalog No. 50-222-3588