Learn More
ACROBiosystems Human NCAM-1 / CD56 Protein, Fc Tag (MALS verified)
Catalog No. 502055065
Encompass_Preferred
Recombinant Protein;1MG;Human NCAM-1, Fc Tag (NC1-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NCAM-1, Fc Tag (Cat. No. NC1-H5253) at 1 μg/mL (100 μL/well) can bind Anti-NCAM-1 Antibody, Human IgG1 with a linear range of 0.3-10 ng/mL (QC tested).
Catalog No. 50-205-5065 Supplier ACROBiosystems Supplier No. NC1H52531MG
Recombinant Protein;1MG;Human NCAM-1, Fc Tag (NC1-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NCAM-1, Fc Tag (Cat. No. NC1-H5253) at 1 μg/mL (100 μL/well) can bind Anti-NCAM-1 Antibody, Human IgG1 with a linear range of 0.3-10 ng/mL (QC tested).