Learn More
Sino Biological Recombinant Human KIR3DL1 Protein (ECD, hFc Tag), HPLC-verified
Catalog No. 502993617
Encompass_Preferred
A DNA sequence encoding the Human KIR3DL1 (P43629) (Met1-His340) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3617 Supplier Sino Biological Supplier No. 12242H02H1
A DNA sequence encoding the Human KIR3DL1 (P43629) (Met1-His340) was expressed with the Fc region of human IgG1 at the C-terminus.