Learn More
Sino Biological Anti-NCAM-1/CD56 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10673MM01A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NCAM1 / CD56 (rh NCAM1 / CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-8437