Learn More
Sino Biological Recombinant Anti-NCAM-1/CD56 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10673R201P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human NCAM1 / CD56 (rh NCAM1 / CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1709