Learn More
Sino Biological Recombinant Human NCAM-1/CD56 Protein (ECD, His & AVI Tag), HPLC-verified, Biotinylated
Supplier:  Sino Biological 10673H27HB100
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met1-Pro603) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5227