Learn More
Sino Biological Anti-NCAPH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100865T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NCAPH, and purified by antigen affinity chromatography.
Catalog No. 50-241-2345