Learn More
Sino Biological Recombinant Human NKp44/NCR2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11550H08HB20
Encompass_Preferred
A DNA sequence encoding the human NCR2 (NP_004819.2) extracellular domain (Met1-Pro190) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4693