Learn More
Sino Biological Recombinant Human NKp30/NCR3 Protein (His Tag), HPLC-verified 500 µg
Supplier:  Sino Biological 10480H08H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 135) of human NCR3 (NP_667341.1) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7175