Learn More
Sino Biological Human NCR3LG1 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 16140H08HB20
Encompass_Preferred
A DNA sequence encoding the human NCR3LG1 (NP_001189368.1) (Met1-Ser262) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-237-3318