Learn More
Sino Biological Human NCSTN Protein 50ug (Sino Biological)
Supplier:  Sino Biological 11183H08H50
Encompass_Preferred
A DNA sequence encoding the human NCSTN (NP_056146.1) extracellular domain (Met 1-Glu 669) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3216